Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172578.2 | NP_766166.2 | 2 | 623 | ||
GenBank mRNA | AK035117.1 | - | 2 | 625 | ||
AK036395.1 | - | 2 | 633 | |||
AK053496.1 | - | 2 | 633 | |||
AK162657.1 | - | 2 | 647 | |||
BC042728.1 | - | 2 | 641 | |||
BC052175.1 | - | 2 | 623 | 6394068 |