Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001148.4 | NP_001139.3 | 1 | 173 | ||
NM_020977.3 | NP_066187.2 | 1 | 173 | |||
XM_005262945.2 | XP_005263002.1 | 1 | 271 | |||
XM_017008068.1 | XP_016863557.1 | 1 | 1236 | |||
XM_017008070.1 | XP_016863559.1 | 1 | 1235 | |||
XM_017008073.1 | XP_016863562.1 | 1 | 271 | |||
XM_017008076.1 | XP_016863565.1 | 1 | 271 | |||
XM_017008078.1 | XP_016863567.1 | 1 | 271 | |||
XM_017008081.1 | XP_016863570.1 | 1 | 271 | |||
XM_017008084.1 | XP_016863573.1 | 1 | 271 | |||
XM_017008085.1 | XP_016863574.1 | 1 | 271 | |||
XM_017008086.1 | XP_016863575.1 | 1 | 271 | |||
XM_017008091.1 | XP_016863580.1 | 1 | 271 | |||
XM_017008094.1 | XP_016863583.1 | 1 | 271 | |||
XM_017008101.1 | XP_016863590.1 | 1 | 271 | |||
XM_017008103.1 | XP_016863592.1 | 1 | 271 | |||
XM_017008106.1 | XP_016863595.1 | 1 | 271 | |||
XM_017008108.1 | XP_016863597.1 | 1 | 271 | |||
GenBank mRNA | AK299767.1 | - | 1 | 152 | ||
AK307601.1 | - | 1 | 124 | |||
X56958.2 | - | 1 | 88 | |||
Z26634.2 | - | 1 | 73 |