Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161.4 | NP_001152.1 | 4 | 456 | ||
NM_001244390.1 | NP_001231319.1 | 2 | 208 | |||
NM_147172.2 | NP_671701.1 | 4 | 385 | |||
NM_147173.2 | NP_671702.1 | 3 | 343 | |||
GenBank mRNA | AA310093.1 | - | 2 | 147 | ||
BC004926.2 | - | 2 | 253 | 3623134 | ||
BG622439.1 | - | 4 | 339 | |||
BG825757.1 | - | 4 | 382 | |||
BM802700.1 | - | 4 | 388 | |||
DN992111.1 | - | 3 | 273 | |||
U30313.1 | - | 3 | 272 |