Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172599.4 | NP_766187.1 | 10 | 1865 | ||
GenBank mRNA | AK029330.1 | - | 10 | 1845 | ||
AK029967.1 | - | 10 | 1846 | |||
AK173037.1 | - | 4 | 919 | |||
BC025038.1 | - | 3 | 856 | |||
BC057360.1 | - | 9 | 1585 | |||
BC090995.1 | - | 10 | 1845 | 6754513 |