Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172712.2 | NP_766300.1 | 4, 5 | 263 | ||
GenBank mRNA | AK029769.1 | - | 4, 5 | 293 | ||
AK049603.1 | - | 4, 5 | 301 | |||
AK053583.1 | - | 4, 5 | 260 | |||
AK086539.1 | - | 4, 5 | 281 | |||
BC063048.1 | - | 4, 5 | 263 | 6825308 |