siRNA Details
Gene Symbol: Cdcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172873.3 | NP_766461.1 | 3 | 1527 | ||
GenBank mRNA | AK050720.1 | - | 3 | 1528 |

More Information
Other Species Targeted

Panther Classification:
