Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199224.1 | NP_001186153.1 | 10 | 1037 | ||
GenBank mRNA | AK052433.1 | - | 4 | 523 | ||
DQ867033.1 | - | 4 | 523 | |||
FN429927.1 | - | 10 | 1037 |