siRNA Details
Gene Symbol: Tox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172913.3 | NP_766501.2 | 3 | 667 | ||
GenBank mRNA | AK082602.1 | - | 3 | 665 | ||
BC052044.1 | - | 3 | 393 | 5708447 | ||
EF410108.1 | - | 531 |

More Information
Other Species Targeted

Panther Classification:
