Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130529.2 | NP_001124001.1 | 7 | 1386 | ||
NM_001291014.1 | NP_001277943.1 | 7 | 1140 | |||
NM_172941.4 | NP_766529.3 | 7 | 1433 | |||
GenBank mRNA | AK051711.1 | - | 7 | 722 | ||
AK139346.1 | - | 7 | 1676 | |||
AK155196.1 | - | 7 | 963 | |||
AK155490.1 | - | 7 | 1015 | |||
AK158448.1 | - | 7 | 973 | |||
AK170924.1 | - | 7 | 963 | |||
AK170989.1 | - | 7 | 968 | |||
AK171190.1 | - | 7 | 1015 | |||
AY302242.1 | - | 6 | 914 | |||
BC040205.1 | - | 6 | 918 | 4947290 |