Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NLGN3 SIRNA Assays ›
Gene Symbol:
Nlgn3
Gene Name:
neuroligin 3
Gene Aliases:
A230085M13Rik, HNL3, NL3
Chromosome Location:
Chr.X: 101299179 - 101321350 on Build GRCm38
UniGene ID:
Mm.121508
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
95355
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nlgn3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_172932.4
NP_766520.2
4
881
GenBank mRNA
AK039018.1
-
4
851
BC069040.1
-
4
1260
BC150773.1
-
3
702
9087684
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_134336, XM_006257054, XM_008773251, XM_006257055, U41663.
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
regulation of respiratory gaseous exchange by neurological system process
receptor-mediated endocytosis
cell adhesion
neuron cell-cell adhesion
synapse assembly
learning
visual learning
adult behavior
social behavior
axon extension
oligodendrocyte differentiation
modulation of synaptic transmission
synapse organization
positive regulation of synapse assembly
regulation of synaptic transmission, glutamatergic
positive regulation of synaptic transmission, glutamatergic
rhythmic synaptic transmission
excitatory postsynaptic potential
inhibitory postsynaptic potential
positive regulation of dendritic spine development
regulation of dendritic spine morphogenesis
vocalization behavior
postsynaptic membrane assembly
presynaptic membrane assembly
positive regulation of inhibitory postsynaptic potential
regulation of long-term synaptic potentiation
positive regulation of protein localization to synapse
regulation of N-methyl-D-aspartate selective glutamate receptor activity
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
regulation of terminal button organization
positive regulation of excitatory postsynaptic potential
positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
receptor activity
neurexin family protein binding
cell adhesion molecule binding
carboxylic ester hydrolase activity
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0