Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173014.1 | NP_766602.1 | 7, 8 | 900 | ||
GenBank mRNA | AB244716.1 | - | 7, 8 | 795 | ||
AK039431.1 | - | 7, 8 | 900 | |||
AK183634.1 | - | 5, 6 | 294 | |||
AK192887.1 | - | 4, 5 | 262 | |||
BC144805.1 | - | 6, 7 | 794 | |||
BC144806.1 | - | 5, 6 | 735 | 9053354 | ||
BC150666.1 | - | 5, 6 | 735 | 9087577 | ||
EF442783.1 | - | 2, 3 | 155 | |||
EF442784.1 | - | 3, 4 | 214 |