Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172951.3 | NP_766539.2 | 14 | 1396 | ||
GenBank mRNA | AF367760.1 | - | 14 | 1251 | ||
AK008673.1 | - | 14 | 1306 | |||
AK035310.1 | - | 14 | 1350 | |||
AK087560.1 | - | 14 | 1346 | |||
AK137345.1 | - | 14 | 1356 | |||
BC058204.1 | - | 13 | 1158 | |||
BC138956.1 | - | 14 | 1310 | 8861978 | ||
BC145476.1 | - | 14 | 1305 |