Search
Search

Tbcel| Gene Symbol: Tbcel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173038.3 | NP_766626.1 | 8 | 4901 | ||
| GenBank mRNA | AK046441.1 | - | 1 | 2306 | ||
| AK077017.1 | - | 8 | 4903 | |||
| AK160836.1 | - | 1 | 2865 | |||
| BC047462.1 | - | 8 | 4766 | |||