Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173404.3 | NP_775580.1 | 1, 2 | 992 | ||
GenBank mRNA | AK033644.1 | - | 1, 2 | 630 | ||
AK035299.1 | - | 1, 2 | 1018 | |||
AK035393.1 | - | 1, 2 | 981 | |||
AK037225.1 | - | 1, 2 | 973 | |||
AK040389.1 | - | 1, 2 | 993 | |||
BC117749.2 | - | 1, 2 | 605 | 40090489 |