Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173405.2 | NP_775581.1 | 7 | 1272 | ||
GenBank mRNA | AJ879912.1 | - | 5 | 913 | ||
AK078332.1 | - | 7 | 1272 | |||
AK155136.1 | - | 1159 | ||||
AK170176.1 | - | 6 | 1246 | |||
BC046428.1 | - | 2 | 240 | |||
BC129841.1 | - | 5 | 1055 | 40090727 | ||
BC129842.1 | - | 5 | 1007 | 40090729 |