Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAV3 SIRNA Assays ›
          Gene Symbol:
          CAV3
          Gene Name:
          caveolin 3
          Gene Aliases:
          LGMD1C, LQT9, VIP-21, VIP21
          Chromosome Location:
          Chr.3: 8733800 - 8746765 on Build GRCh38
          UniGene ID:
          Hs.98303
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 9576

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAV3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001234.4 NP_001225.1 1, 2 199
          NM_033337.2 NP_203123.1 1, 2 199
          GenBank mRNA AF036365.1 - 1, 2 169
          AF043101.1 - 1, 2 119
          AK291892.1 - 1, 2 199
          BC069368.1 - 1, 2 172 7262168
          BC102033.1 - 1, 2 159 40033505
          BC102036.1 - 1, 2 159 40033114
          BC102037.1 - 1, 2 159 40033118
          Y14747.1 - 1, 2 133

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane repair
          regulation of heart rate
          negative regulation of protein kinase activity
          triglyceride metabolic process
          endocytosis
          muscle contraction
          plasma membrane organization
          actin filament organization
          positive regulation of cytosolic calcium ion concentration
          muscle organ development
          myoblast fusion
          regulation of heart contraction
          protein localization
          positive regulation of cell proliferation
          negative regulation of cardiac muscle hypertrophy
          positive regulation of myotube differentiation
          regulation of skeletal muscle contraction
          cell growth
          regulation of transforming growth factor beta receptor signaling pathway
          cell differentiation
          positive regulation of microtubule polymerization
          cytoplasmic microtubule organization
          membrane raft organization
          T-tubule organization
          detection of muscle stretch
          regulation of signal transduction by receptor internalization
          regulation of membrane potential
          glucose homeostasis
          cholesterol homeostasis
          negative regulation of MAP kinase activity
          negative regulation of MAPK cascade
          negative regulation of cell size
          muscle cell cellular homeostasis
          negative regulation of nitric-oxide synthase activity
          regulation of nerve growth factor receptor activity
          nucleus localization
          regulation of protein kinase B signaling
          negative regulation of calcium ion transport
          cardiac muscle cell development
          regulation of cardiac muscle contraction
          negative regulation of sarcomere organization
          regulation of ventricular cardiac muscle cell membrane repolarization
          heart trabecula formation
          regulation of ventricular cardiac muscle cell membrane depolarization
          regulation of branching involved in mammary gland duct morphogenesis
          negative regulation of cell growth involved in cardiac muscle cell development
          caveola assembly
          protein localization to plasma membrane
          ventricular cardiac muscle cell action potential
          establishment of protein localization to plasma membrane
          regulation of calcium ion import
          regulation of p38MAPK cascade
          regulation of membrane depolarization during cardiac muscle cell action potential
          negative regulation of membrane depolarization during cardiac muscle cell action potential
          negative regulation of potassium ion transmembrane transporter activity
          regulation of calcium ion transmembrane transporter activity
          negative regulation of potassium ion transmembrane transport
          negative regulation of protein localization to cell surface
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          regulation of sodium ion transmembrane transporter activity
          positive regulation of caveolin-mediated endocytosis
          calcium channel regulator activity
          protein binding
          protein C-terminus binding
          sodium channel regulator activity
          potassium channel inhibitor activity
          protein complex binding
          protein complex scaffold
          alpha-tubulin binding
          ion channel binding
          nitric-oxide synthase binding
          connexin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0