Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ENTPD6 SIRNA Assays ›
Gene Symbol:
ENTPD6
Gene Name:
ectonucleoside triphosphate diphosphohydrolase 6 (putative)
Gene Aliases:
CD39L2, IL-6SAG, IL6ST2, NTPDase-6, dJ738P15.3
Chromosome Location:
Chr.20: 25195670 - 25228075 on Build GRCh38
UniGene ID:
Hs.500375
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9582
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ENTPD6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114089.3
NP_001107561.1
3
573
NM_001247.4
NP_001238.2
4
642
NM_001317941.2
NP_001304870.1
4
639
NM_001322378.1
NP_001309307.1
4
786
NM_001322379.1
NP_001309308.1
2
251
NM_001322380.1
NP_001309309.1
2
251
NM_001322381.1
NP_001309310.1
2
251
NM_001322382.1
NP_001309311.1
2
251
NM_001322383.1
NP_001309312.1
4
639
NM_001322384.1
NP_001309313.1
2
251
NM_001322385.1
NP_001309314.1
3
573
NM_001322386.1
NP_001309315.1
3
573
NM_001322387.1
NP_001309316.1
4
642
NM_001322388.1
NP_001309317.1
4
642
NM_001322389.1
NP_001309318.1
3
717
NM_001322390.1
NP_001309319.1
5
764
NM_001322391.1
NP_001309320.1
4
642
NM_001322392.1
NP_001309321.1
3
570
NM_001322393.1
NP_001309322.1
3
573
NM_001322394.1
NP_001309323.1
3
570
NM_001322395.1
NP_001309324.1
3
717
NM_001322396.1
NP_001309325.1
4
639
NM_001322397.1
NP_001309326.1
4
642
NM_001322398.1
NP_001309327.1
3
717
XM_006723665.2
XP_006723728.1
5
678
XM_011529396.1
XP_011527698.1
5
682
XM_011529397.1
XP_011527699.1
5
611
XM_017028129.1
XP_016883618.1
3
451
XM_017028130.1
XP_016883619.1
3
451
XM_017028131.1
XP_016883620.1
3
451
XM_017028132.1
XP_016883621.1
4
618
XM_017028133.1
XP_016883622.1
3
421
XM_017028134.1
XP_016883623.1
2
218
GenBank mRNA
AF039916.1
-
4
576
AK074691.1
-
3
422
AK128412.1
-
2
190
AK293256.1
-
3
514
AK297954.1
-
4
482
AK300028.1
-
2
202
AK300514.1
-
2
177
AK302395.1
-
4
490
AK303786.1
-
3
528
AK308626.1
-
3
540
AL834158.1
-
3
488
AY327581.1
-
4
539
BC025980.2
-
4
546
3636163
CR936750.1
-
2
199
CR936765.1
-
4
556
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
nucleotide phosphatase
phosphatase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
response to magnesium ion
response to calcium ion
guanosine-5'-triphosphate,3'-diphosphate diphosphatase activity
nucleoside-triphosphatase activity
uridine-diphosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0