siRNA Details
Gene Symbol: Rbbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175023.3 | NP_778188.1 | 3 | 1675 | ||
GenBank mRNA | AK045635.1 | - | 3 | 1420 | ||
AK082686.1 | - | 1 | 1017 | |||
AK164942.1 | - | 1 | 55 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
