Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK2 SIRNA Assays ›
Gene Symbol:
ANK2
Gene Name:
ankyrin 2, neuronal
Gene Aliases:
ANK-2, LQT4, brank-2
Chromosome Location:
Chr.4: 112706083 - 113383740 on Build GRCh38
UniGene ID:
Hs.620557
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9629
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127493.1
NP_001120965.1
41
4732
NM_001148.4
NP_001139.3
40
10893
NM_020977.3
NP_066187.2
39
4638
XM_005262945.2
XP_005263002.1
39
10892
XM_017008067.1
XP_016863556.1
42
11850
XM_017008068.1
XP_016863557.1
43
12091
XM_017008069.1
XP_016863558.1
41
11825
XM_017008070.1
XP_016863559.1
42
12036
XM_017008071.1
XP_016863560.1
43
11019
XM_017008072.1
XP_016863561.1
42
10995
XM_017008073.1
XP_016863562.1
40
10991
XM_017008074.1
XP_016863563.1
42
10965
XM_017008075.1
XP_016863564.1
41
10929
XM_017008076.1
XP_016863565.1
40
10991
XM_017008077.1
XP_016863566.1
39
10806
XM_017008078.1
XP_016863567.1
39
4736
XM_017008079.1
XP_016863568.1
37
4466
XM_017008080.1
XP_016863569.1
39
4587
XM_017008081.1
XP_016863570.1
38
4637
XM_017008082.1
XP_016863571.1
40
4814
XM_017008083.1
XP_016863572.1
40
4877
XM_017008084.1
XP_016863573.1
40
4772
XM_017008085.1
XP_016863574.1
40
4733
XM_017008086.1
XP_016863575.1
39
4634
XM_017008087.1
XP_016863576.1
41
4913
XM_017008088.1
XP_016863577.1
42
4707
XM_017008089.1
XP_016863578.1
39
4880
XM_017008090.1
XP_016863579.1
40
4877
XM_017008091.1
XP_016863580.1
38
4574
XM_017008092.1
XP_016863581.1
40
4674
XM_017008093.1
XP_016863582.1
41
4671
XM_017008094.1
XP_016863583.1
40
4733
XM_017008095.1
XP_016863584.1
38
4551
XM_017008096.1
XP_016863585.1
42
4707
XM_017008097.1
XP_016863586.1
39
4512
XM_017008098.1
XP_016863587.1
40
4674
XM_017008099.1
XP_016863588.1
41
4671
XM_017008100.1
XP_016863589.1
38
4476
XM_017008101.1
XP_016863590.1
38
4637
XM_017008102.1
XP_016863591.1
37
4452
XM_017008103.1
XP_016863592.1
40
4733
XM_017008104.1
XP_016863593.1
39
4575
XM_017008105.1
XP_016863594.1
38
4551
XM_017008106.1
XP_016863595.1
37
4538
XM_017008107.1
XP_016863596.1
39
4512
XM_017008108.1
XP_016863597.1
37
4436
XM_017008109.1
XP_016863598.1
37
4503
XM_017008110.1
XP_016863599.1
39
4575
XM_017008111.1
XP_016863600.1
38
4551
XM_017008112.1
XP_016863601.1
36
4353
GenBank mRNA
AK294720.1
-
18
2383
AK299893.1
-
12
1480
BC125235.1
-
7
523
BC125236.1
-
7
523
BX537758.1
-
41
4706
BX538132.1
-
12
1542
BX538239.1
-
4
2798
X56958.2
-
39
4553
Z26634.2
-
40
10793
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
atrial septum development
cellular calcium ion homeostasis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
positive regulation of gene expression
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of cardiac muscle contraction by calcium ion signaling
paranodal junction assembly
regulation of protein stability
T-tubule organization
protein localization to organelle
protein localization to cell surface
cellular protein localization
protein localization to M-band
protein localization to T-tubule
positive regulation of potassium ion transport
protein stabilization
regulation of release of sequestered calcium ion into cytosol
response to methylmercury
regulation of calcium ion transport
positive regulation of calcium ion transport
regulation of cardiac muscle contraction
regulation of ventricular cardiac muscle cell membrane repolarization
sarcoplasmic reticulum calcium ion transport
protein localization to endoplasmic reticulum
protein localization to plasma membrane
protein targeting to plasma membrane
regulation of cardiac muscle cell contraction
ventricular cardiac muscle cell action potential
atrial cardiac muscle cell action potential
SA node cell action potential
membrane depolarization during SA node cell action potential
atrial cardiac muscle cell to AV node cell communication
SA node cell to atrial cardiac muscle cell communication
regulation of heart rate by cardiac conduction
regulation of SA node cell action potential
regulation of atrial cardiac muscle cell action potential
positive regulation of potassium ion transmembrane transporter activity
regulation of calcium ion transmembrane transporter activity
positive regulation of calcium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
protein kinase binding
spectrin binding
protein binding, bridging
ion channel binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0