Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMK2B SIRNA Assays ›
Gene Symbol:
CAMK2B
Gene Name:
calcium/calmodulin dependent protein kinase II beta
Gene Aliases:
CAM2, CAMK2, CAMKB
Chromosome Location:
Chr.7: 44217150 - 44325994 on Build GRCh38
UniGene ID:
Hs.351887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9662
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMK2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220.4
NP_001211.3
21
1826
NM_001293170.1
NP_001280099.1
18
1454
NM_172078.2
NP_742075.1
18
1454
NM_172079.2
NP_742076.1
17
1382
NM_172080.2
NP_742077.1
17
1379
NM_172081.2
NP_742078.1
16
1337
NM_172082.2
NP_742079.1
17
1382
NM_172083.2
NP_742080.1
15
1265
NM_172084.2
NP_742081.1
13
1175
XM_005249862.3
XP_005249919.1
17
1285
XM_005249864.3
XP_005249921.1
16
1186
XM_006715781.2
XP_006715844.1
19
1582
XM_006715784.2
XP_006715847.1
17
1258
XM_011515547.1
XP_011513849.1
22
1831
XM_011515549.1
XP_011513851.1
21
1786
XM_011515550.1
XP_011513852.1
21
1759
XM_011515551.1
XP_011513853.1
21
1759
XM_011515552.1
XP_011513854.1
21
1756
XM_011515553.1
XP_011513855.1
21
1717
XM_011515554.1
XP_011513856.1
20
1711
XM_011515555.1
XP_011513857.1
21
1702
XM_011515556.1
XP_011513858.1
20
1687
XM_011515557.1
XP_011513859.1
20
1684
XM_011515558.2
XP_011513860.1
23
1960
XM_011515559.1
XP_011513861.1
22
1740
XM_017012660.1
XP_016868149.1
20
1714
XM_017012661.1
XP_016868150.1
20
1672
XM_017012662.1
XP_016868151.1
19
1513
XM_017012663.1
XP_016868152.1
21
1668
XM_017012664.1
XP_016868153.1
16
1210
XM_017012665.1
XP_016868154.1
16
1183
XM_017012666.1
XP_016868155.1
18
1239
XM_017012667.1
XP_016868156.1
17
1194
XM_017012668.1
XP_016868157.1
17
1167
XM_017012669.1
XP_016868158.1
17
1167
XM_017012670.1
XP_016868159.1
16
1122
GenBank mRNA
AF078803.1
-
18
1247
AF081572.1
-
16
1130
AF081924.1
-
17
1175
AF083419.1
-
17
1172
AF112471.1
-
17
1215
AF112472.1
-
18
1290
AF140350.1
-
13
968
AJ252236.1
-
16
1214
AJ252237.1
-
15
1065
AJ252238.1
-
17
1172
AK091991.1
-
3
1701
AK222703.1
-
18
1278
AK290148.1
-
18
1319
AK294138.1
-
16
1177
AK315663.1
-
16
1244
BC019070.2
-
16
1331
5014712
U23460.1
-
21
1685
U50358.1
-
5
224
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein phosphorylation
signal transduction
positive regulation of neuron projection development
regulation of skeletal muscle adaptation
positive regulation of phospholipase A2 activity
positive regulation of GTPase activity
protein autophosphorylation
regulation of long-term neuronal synaptic plasticity
cell projection morphogenesis
regulation of synapse structural plasticity
regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
regulation of dendritic spine development
positive regulation of dendritic spine morphogenesis
positive regulation of synapse maturation
regulation of cellular response to heat
actin binding
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
protein kinase binding
protein homodimerization activity
phospholipase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0