Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clvs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175448.3 | NP_780657.1 | 4 | 1188 | ||
GenBank mRNA | AK039308.1 | - | 4 | 1189 | ||
AK049896.1 | - | 4 | 749 | |||
AK049988.1 | - | 4 | 757 | |||
AK139409.1 | - | 4 | 1263 | |||
AK139868.1 | - | 4 | 1241 | |||
AK169009.1 | - | 4 | 1358 | |||
BC058539.1 | - | 4 | 1195 | 5690124 |