Gene Symbol: Pnma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175498.4 | NP_780707.1 | 3 | 770 | ||
GenBank mRNA | AK043718.1 | - | 3 | 718 | ||
AK043910.1 | - | 2 | 640 | |||
AK220357.1 | - | 725 | ||||
BC065116.1 | - | 2 | 601 | 30618233 |