Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175550.3 | NP_780759.2 | 2 | 222 | ||
GenBank mRNA | AK036944.1 | - | 2 | 205 | ||
AK039257.1 | - | 2 | 210 | |||
AK143351.1 | - | 2 | 212 | |||
AK144636.1 | - | 2 | 202 | |||
AK163701.1 | - | 2 | 213 | |||
BY037321.1 | - | 2 | 223 |