Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175642.4 | NP_783573.4 | 2 | 497 | ||
GenBank mRNA | AY168406.1 | - | 1 | 236 | ||
BC099951.1 | - | 2 | 495 | 30098919 |
Set Membership: |
G-protein coupled receptor |