Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029654.4 | NP_083930.5 | 3 | 806 | ||
GenBank mRNA | AK032840.1 | - | 1 | 72 | ||
AK083213.1 | - | 3 | 766 | |||
AK085790.1 | - | 3 | 753 | |||
AK086802.1 | - | 3 | 807 | |||
BC062182.1 | - | 3 | 776 | 6505402 |