Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005510.2 | NP_001005510.2 | 3 | 304 | ||
GenBank mRNA | AF435012.1 | - | 3 | 288 | ||
AK052521.1 | - | 3 | 273 | |||
AK078542.1 | - | 3 | 305 | |||
AK084394.1 | - | 4 | 484 | |||
AK084811.1 | - | 3 | 302 | |||
AK145252.1 | - | 3 | 305 |