Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018729.2 | NP_061199.2 | 9 | 2181 | ||
GenBank mRNA | AK033178.1 | - | 4 | 1164 | ||
AK155548.1 | - | 9 | 2128 | |||
AK172371.1 | - | 1 | 270 |