Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005510.2 | NP_001005510.2 | 5 | 467 | ||
GenBank mRNA | AK052521.1 | - | 5 | 436 | ||
AK078542.1 | - | 5 | 468 | |||
AK084394.1 | - | 6 | 647 | |||
AK084811.1 | - | 5 | 467 | |||
AK145252.1 | - | 5 | 468 |