Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Relt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177073.6 | NP_796047.2 | 4 | 285 | ||
GenBank mRNA | AK042363.1 | - | 4 | 280 | ||
AK049073.1 | - | 4 | 286 | |||
AK088621.1 | - | 4 | 268 | |||
BC100310.1 | - | 4 | 275 | 30839974 | ||
BC131929.1 | - | 4 | 196 | 40130206 | ||
BC131931.1 | - | 4 | 196 | 40130208 | ||
BC145200.1 | - | 4 | 196 | 9053751 |