Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205227.1 | NP_001192156.1 | 2, 3 | 207 | ||
NM_177216.4 | NP_796190.1 | 2, 3 | 207 | |||
GenBank mRNA | AK085272.1 | - | 2, 3 | 172 | ||
AK147952.1 | - | 2, 3 | 208 | |||
BC107238.1 | - | 2, 3 | 151 | 40052160 | ||
BC107239.1 | - | 2, 3 | 151 | 40052161 |