Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252313.1 | NP_001239242.1 | 2, 3 | 323 | ||
NM_011308.3 | NP_035438.3 | 2, 3 | 323 | |||
NM_177229.3 | NP_796203.1 | 3, 4 | 508 | |||
GenBank mRNA | AK035813.1 | - | 3, 4 | 500 | ||
AK077420.1 | - | 3, 4 | 509 | |||
BC026623.1 | - | 3, 4 | 694 | |||
BC029627.1 | - | 3, 4 | 486 | |||
BC062162.1 | - | 3, 4 | 486 | |||
BC086657.1 | - | 2, 3 | 289 | 30545994 |