Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLG2 SIRNA Assays ›
          Gene Symbol:
          DLG2
          Gene Name:
          discs large MAGUK scaffold protein 2
          Gene Aliases:
          PPP1R58, PSD-93, PSD93, chapsyn-110
          Chromosome Location:
          Chr.11: 83455009 - 85628534 on Build GRCh38
          UniGene ID:
          Hs.367656
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 9830

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DLG2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142699.1 NP_001136171.1 10, 11 1069
          NM_001142700.1 NP_001136172.1 3, 4 314
          NM_001206769.1 NP_001193698.1 4, 5 753
          NM_001300983.1 NP_001287912.1 5, 6 745
          NM_001364.3 NP_001355.2 5, 6 745
          XM_005273810.4 XP_005273867.1 4, 5 388
          XM_005273811.4 XP_005273868.1 4, 5 388
          XM_011544778.2 XP_011543080.1 11, 12 2360
          XM_011544780.2 XP_011543082.1 11, 12 2360
          XM_011544782.2 XP_011543084.1 6, 7 831
          XM_011544788.2 XP_011543090.1 4, 5 388
          XM_017017254.1 XP_016872743.1 11, 12 2360
          XM_017017255.1 XP_016872744.1 10, 11 2321
          XM_017017256.1 XP_016872745.1 10, 11 1850
          XM_017017257.1 XP_016872746.1 9, 10 2782
          XM_017017258.1 XP_016872747.1 8, 9 1958
          XM_017017259.1 XP_016872748.1 9, 10 1775
          XM_017017260.1 XP_016872749.1 8, 9 2707
          XM_017017261.1 XP_016872750.1 11, 12 2360
          XM_017017262.1 XP_016872751.1 4, 5 813
          XM_017017263.1 XP_016872752.1 6, 7 831
          XM_017017264.1 XP_016872753.1 5, 6 1008
          XM_017017265.1 XP_016872754.1 5, 6 1005
          XM_017017266.1 XP_016872755.1 4, 5 811
          XM_017017267.1 XP_016872756.1 5, 6 913
          XM_017017268.1 XP_016872757.1 5, 6 734
          XM_017017269.1 XP_016872758.1 5, 6 1435
          XM_017017270.1 XP_016872759.1 6, 7 831
          XM_017017271.1 XP_016872760.1 5, 6 1008
          XM_017017272.1 XP_016872761.1 5, 6 1005
          XM_017017273.1 XP_016872762.1 4, 5 820
          XM_017017274.1 XP_016872763.1 5, 6 913
          XM_017017275.1 XP_016872764.1 4, 5 812
          XM_017017276.1 XP_016872765.1 5, 6 1433
          XM_017017277.1 XP_016872766.1 4, 5 657
          XM_017017278.1 XP_016872767.1 4, 5 813
          XM_017017279.1 XP_016872768.1 6, 7 831
          XM_017017280.1 XP_016872769.1 6, 7 1508
          XM_017017281.1 XP_016872770.1 5, 6 1008
          XM_017017282.1 XP_016872771.1 4, 5 388
          XM_017017283.1 XP_016872772.1 4, 5 657
          XM_017017284.1 XP_016872773.1 3, 4 233
          XM_017017285.1 XP_016872774.1 5, 6 1435
          GenBank mRNA AB209252.1 - 5, 6 720
          AK126776.1 - 3, 4 314
          AK127150.1 - 4, 5 751
          AK294375.1 - 4, 5 276
          AK295094.1 - 5, 6 745
          AK295624.1 - 4, 5 368
          AK299797.1 - 5, 6 503
          CR749820.1 - 10, 11 1069
          KJ535093.1 - 10, 11 952
          U32376.1 - 5, 6 718

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          nervous system development
          negative regulation of phosphatase activity
          sensory perception of pain
          receptor clustering
          establishment or maintenance of epithelial cell apical/basal polarity
          GMP metabolic process
          GDP metabolic process
          receptor localization to synapse
          guanylate kinase activity
          protein binding
          kinase binding
          ionotropic glutamate receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0