siRNA Details
Gene Symbol: Dync2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029851.2 | NP_084127.2 | 20 | 2959 | ||
GenBank mRNA | AK046538.1 | - | 18 | 3728 | ||
AY452064.1 | - | 3251 | ||||
DQ104402.1 | - | 20 | 2959 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
