Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290374.1 | NP_001277303.1 | 2 | 656 | ||
NM_001290375.1 | NP_001277304.1 | 2 | 656 | |||
NM_001290376.1 | NP_001277305.1 | 2 | 172 | |||
NM_177343.4 | NP_796317.2 | 2 | 656 | |||
GenBank mRNA | AK052401.1 | - | 2 | 77 | ||
AK053173.1 | - | 2 | 410 | |||
AK141415.1 | - | 2 | 397 | |||
AK147616.1 | - | 2 | 657 | |||
AK147657.1 | - | 2 | 367 | |||
AK154434.1 | - | 2 | 245 | |||
AK162754.1 | - | 2 | 371 | |||
AK170123.1 | - | 2 | 415 | |||
AK170777.1 | - | 2 | 421 | |||
AY273822.1 | - | 2 | 172 | |||
BC141413.1 | - | 2 | 156 | 9056026 | ||
BC141414.1 | - | 2 | 156 | 9056027 |