Search
Search

DYNC1I2| Gene Symbol: DYNC1I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271785.1 | NP_001258714.1 | 7 | 619 | ||
| NM_001271786.1 | NP_001258715.1 | 5 | 612 | |||
| NM_001271787.1 | NP_001258716.1 | 5 | 595 | |||
| NM_001271788.1 | NP_001258717.1 | 5 | 558 | |||
| NM_001271789.1 | NP_001258718.1 | 5 | 541 | |||
| NM_001271790.1 | NP_001258719.1 | 5 | 558 | |||
| NM_001320882.1 | NP_001307811.1 | 6 | 618 | |||
| NM_001320883.1 | NP_001307812.1 | 6 | 601 | |||
| NM_001320884.1 | NP_001307813.1 | 5 | 541 | |||
| NM_001378.2 | NP_001369.1 | 7 | 636 | |||
| XM_006712347.3 | XP_006712410.1 | 6 | 542 | |||
| XM_017003525.1 | XP_016859014.1 | 6 | 577 | |||
| GenBank mRNA | AB209014.1 | - | 3 | 561 | ||
| AF134477.1 | - | 5 | 430 | |||
| AF250307.1 | - | 5 | 450 | |||
| AK055491.1 | - | 7 | 573 | |||
| AK096579.1 | - | 7 | 537 | |||
| AK225759.1 | - | 5 | 504 | |||
| AK298561.1 | - | 5 | 455 | |||
| AK308008.1 | - | 5 | 413 | |||
| AK316119.1 | - | 5 | 475 | |||
| AY037160.1 | - | 5 | 466 | |||
| BC015038.2 | - | 5 | 402 | 3921299 | ||
| BC091498.1 | - | 5 | 486 | 30386904 | ||
| BC109375.1 | - | 5 | 443 | 4608676 | ||
| BP355434.1 | - | 5 | 542 | |||
| BT007130.1 | - | 4 | 330 | |||
| BX537412.1 | - | 5 | 498 | |||
| U39575.1 | - | 3 | 251 | |||