Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt61a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099792.1 | NP_001093262.1 | 4 | 1109 | ||
NM_001099793.1 | NP_001093263.1 | 4 | 970 | |||
NM_177374.4 | NP_796348.2 | 3 | 1411 | |||
GenBank mRNA | AK032824.1 | - | 3 | 1374 | ||
AK049138.1 | - | 3 | 1382 | |||
BC051186.1 | - | 3 | 1365 | 6468754 |