Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001398.2 | NP_001389.2 | 2, 3 | 342 | ||
XM_017026448.1 | XP_016881937.1 | 2, 3 | 292 | |||
GenBank mRNA | AK291860.1 | - | 2, 3 | 277 | ||
AK301211.1 | - | 2, 3 | 274 | |||
BC011792.2 | - | 2, 3 | 259 | 4300082 | ||
BC017408.1 | - | 2, 3 | 283 | 4306900 | ||
BG527600.1 | - | 2, 3 | 343 | |||
U16660.1 | - | 2, 3 | 284 |