Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177606.4 | NP_808274.2 | 13 | 2250 | ||
GenBank mRNA | AK029252.1 | - | 13 | 2252 | ||
AK173337.1 | - | 10 | 1905 | |||
BC125583.1 | - | 13 | 2215 | 40130098 | ||
BC137804.1 | - | 13 | 2212 | 8860823 |