Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctso | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177662.2 | NP_808330.1 | 3 | 374 | ||
GenBank mRNA | AK034490.1 | - | 3 | 374 | ||
AK040071.1 | - | 3 | 348 | |||
AK049470.1 | - | 3 | 347 | |||
AK143683.1 | - | 3 | 390 | |||
AK165930.1 | - | 3 | 336 | |||
AK166103.1 | - | 3 | 343 | |||
BC044664.1 | - | 3 | 266 | |||
BC098219.1 | - | 3 | 348 | 1066724 |