Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177766.3 | NP_808434.2 | 5 | 932 | ||
GenBank mRNA | AK031600.1 | - | 5 | 680 | ||
AK200930.1 | - | 2 | 195 | |||
BC115553.1 | - | 5 | 591 | 40101801 | ||
BC115554.1 | - | 5 | 591 | 40101802 |