Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRAK4 SIRNA Assays ›
Gene Symbol:
IRAK4
Gene Name:
interleukin 1 receptor associated kinase 4
Gene Aliases:
IPD1, IRAK-4, NY-REN-64, REN64
Chromosome Location:
Chr.12: 43758913 - 43789543 on Build GRCh38
UniGene ID:
Hs.138499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
993
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRAK4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114182.2
NP_001107654.1
3
229
NM_001145256.1
NP_001138728.1
3
229
NM_001145257.1
NP_001138729.1
2
181
NM_016123.3
NP_057207.2
2
181
XM_005268943.3
XP_005269000.1
2
286
XM_005268944.4
XP_005269001.1
2
250
XM_005268945.4
XP_005269002.1
2
494
XM_005268947.4
XP_005269004.1
2
173
XM_005268948.2
XP_005269005.1
3
213
XM_005268949.2
XP_005269006.1
2
286
XM_006719438.3
XP_006719501.1
3
542
XM_006719439.2
XP_006719502.1
3
793
XM_006719440.2
XP_006719503.1
2
144
XM_006719441.2
XP_006719504.1
2
175
XM_006719442.2
XP_006719505.1
3
179
XM_006719443.3
XP_006719506.1
2
322
XM_011538431.2
XP_011536733.1
3
298
XM_011538432.1
XP_011536734.1
3
334
XM_011538433.2
XP_011536735.1
2
913
XM_017019390.1
XP_016874879.1
4
415
XM_017019391.1
XP_016874880.1
2
313
GenBank mRNA
AF155118.1
-
2
148
AF445802.1
-
1
99
AK000528.1
-
2
128
AK027301.2
-
3
822
AK299944.1
-
2
178
AK314288.1
-
2
170
AY283670.1
-
1
99
AY283671.1
-
1
99
AY340962.1
-
2
164
AY340963.1
-
3
212
AY340964.1
-
2
164
AY340965.1
-
3
212
AY340966.1
-
2
164
AY340967.1
-
3
212
AY730682.1
-
1
99
BC013316.1
-
2
169
4287014
DA290809.1
-
2
175
KU178631.1
-
1
99
KU178632.1
-
1
99
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
toll-like receptor signaling pathway
neutrophil mediated immunity
MyD88-dependent toll-like receptor signaling pathway
JNK cascade
cytokine-mediated signaling pathway
toll-like receptor 9 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of smooth muscle cell proliferation
neutrophil migration
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
interleukin-1 receptor binding
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0