Search
Search

Ints8| Gene Symbol: Ints8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159595.1 | NP_001153067.1 | 2 | 328 | ||
| NM_178112.5 | NP_835213.3 | 2 | 328 | |||
| GenBank mRNA | AK049513.1 | - | 2 | 299 | ||
| AK140829.1 | - | 2 | 323 | |||
| AK169552.1 | - | 2 | 297 | |||
| BC049092.1 | - | 2 | 258 | 6402150 | ||
| BC057618.1 | - | 2 | 258 | 6402150 | ||
| CJ134643.1 | - | 2 | 331 | |||