Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178347.4 | NP_848124.1 | 3 | 283 | ||
GenBank mRNA | AK031459.1 | - | 3 | 266 | ||
AK038523.1 | - | 3 | 266 | |||
AK047789.1 | - | 3 | 266 | |||
AK048246.1 | - | 3 | 266 | |||
AK052567.1 | - | 3 | 283 | |||
AK075948.1 | - | 3 | 284 | |||
AK079681.1 | - | 2 | 350 | |||
AK081461.1 | - | 3 | 284 | |||
BC059013.1 | - | 3 | 252 | 6813730 |