Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FHL1 SIRNA Assays ›
Gene Symbol:
FHL1
Gene Name:
four and a half LIM domains 1
Gene Aliases:
FHL-1, FHL1A, FHL1B, FLH1A, KYOT, RBMX1A, RBMX1B, SLIM, SLIM-1, SLIM1, SLIMMER, XMPMA
Chromosome Location:
Chr.X: 136146702 - 136211359 on Build GRCh38
UniGene ID:
Hs.435369
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9962
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FHL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159699.1
NP_001153171.1
2, 3
429
NM_001159700.1
NP_001153172.1
3, 4
424
NM_001159701.1
NP_001153173.1
2, 3
300
NM_001159702.2
NP_001153174.1
3, 4
493
NM_001159703.1
NP_001153175.1
3, 4
493
NM_001159704.1
NP_001153176.1
2, 3
875
NM_001167819.1
NP_001161291.1
3, 4
361
NM_001449.4
NP_001440.2
3, 4
493
NR_027621.1
3, 4
575
XM_006724743.2
XP_006724806.1
2, 3
297
XM_006724744.2
XP_006724807.1
3, 4
375
XM_006724745.3
XP_006724808.1
3, 4
313
XM_006724746.2
XP_006724809.1
2, 3
248
XM_006724747.2
XP_006724810.1
2, 3
248
XM_011531316.1
XP_011529618.1
2, 3
297
XM_017029357.1
XP_016884846.1
3, 4
375
GenBank mRNA
AF063002.1
-
2, 3
248
AF098518.1
-
2, 3
217
AF220153.1
-
1, 2
164
AK122708.1
-
2, 3
876
AK223299.1
-
1, 2
198
AK289411.1
-
3, 4
575
AK299381.1
-
2, 3
275
AK299450.1
-
2, 3
272
AK300966.1
-
4, 5
300
AK301642.1
-
2, 3
297
AK304680.1
-
3, 4
352
BC010998.2
-
3, 4
372
4039973
BC088369.1
-
2, 3
262
30529295
BE965488.2
-
3, 4
301
CR456974.1
-
1, 2
164
DA497183.1
-
3, 4
493
DA917361.1
-
1, 2
59
DA982372.1
-
3, 4
362
DC363191.1
-
3, 4
424
DC373304.1
-
2, 3
301
DC373386.1
-
2, 3
430
U29538.1
-
1, 2
185
U60115.1
-
2, 3
247
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of membrane depolarization
muscle organ development
organ morphogenesis
negative regulation of G2/M transition of mitotic cell cycle
cell differentiation
negative regulation of cell growth
positive regulation of potassium ion transport
regulation of potassium ion transmembrane transporter activity
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
protein binding
zinc ion binding
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0