Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other THRA SIRNA Assays ›
Gene Symbol:
Thra
Gene Name:
thyroid hormone receptor alpha
Gene Aliases:
6430529J03Rik, AW259572, Erba, Nr1a1, Rvr, T3R[a], T3Ralpha, Thra1, Thra2
Chromosome Location:
Chr.11: 98741873 - 98765111 on Build GRCm38
UniGene ID:
Mm.265917
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
99653
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Thra
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_178060.3
NP_835161.1
9
2175
GenBank mRNA
AK078233.1
-
9
2175
AK165172.1
-
185
Back To Top
More Information
Panther Classification:
Molecular Function -
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
cartilage condensation
ossification
regulation of thyroid hormone mediated signaling pathway
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
learning or memory
regulation of heart contraction
female courtship behavior
response to cold
hormone-mediated signaling pathway
organ morphogenesis
positive regulation of myotube differentiation
negative regulation of RNA polymerase II transcriptional preinitiation complex assembly
erythrocyte differentiation
intracellular receptor signaling pathway
thyroid gland development
regulation of myeloid cell apoptotic process
cytoplasmic sequestering of transcription factor
steroid hormone mediated signaling pathway
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of transcription, DNA-templated
positive regulation of female receptivity
positive regulation of transcription from RNA polymerase II promoter
regulation of lipid catabolic process
Type I pneumocyte differentiation
negative regulation of DNA-templated transcription, initiation
steroid receptor RNA activator RNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
steroid hormone receptor activity
single-stranded RNA binding
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
thyroid hormone receptor activity
protein binding
transcription factor binding
zinc ion binding
TBP-class protein binding
protein domain specific binding
chromatin DNA binding
protein complex binding
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
intronic transcription regulatory region DNA binding
metal ion binding
protein heterodimerization activity
protein dimerization activity
thyroid hormone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0