Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001496.3 | NP_001487.2 | 2 | 429 | ||
GenBank mRNA | AF051767.1 | - | 2 | 289 | ||
AK297693.1 | - | 2 | 383 | |||
AK314022.1 | - | 2 | 330 | |||
AY358997.1 | - | 2 | 326 | |||
AY359037.1 | - | 2 | 430 | |||
BC037951.1 | - | 2 | 314 | 5227725 | ||
FJ200487.1 | - | 2 | 289 |