Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTN1 SIRNA Assays ›
          Gene Symbol:
          ACTN1
          Gene Name:
          actinin alpha 1
          Gene Aliases:
          BDPLT15
          Chromosome Location:
          Chr.14: 68874123 - 68979366 on Build GRCh38
          UniGene ID:
          Hs.235750
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS100130
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACTN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001102.3 NP_001093.1 10 1226
          NM_001130004.1 NP_001123476.1 10 1226
          NM_001130005.1 NP_001123477.1 10 1226
          XM_011537265.2 XP_011535567.1 10 1018
          XM_011537266.2 XP_011535568.1 10 1018
          XM_011537267.2 XP_011535569.1 10 1018
          XM_011537268.2 XP_011535570.1 10 1018
          XM_017021720.1 XP_016877209.1 10 1018
          XM_017021721.1 XP_016877210.1 10 1018
          XM_017021722.1 XP_016877211.1 10 1163
          XM_017021723.1 XP_016877212.1 10 1163
          XM_017021725.1 XP_016877214.1 10 1337
          XM_017021726.1 XP_016877215.1 10 1337
          XM_017021727.1 XP_016877216.1 10 1163
          XM_017021728.1 XP_016877217.1 10 1337
          GenBank mRNA AK225782.1 - 10 1123
          AK295099.1 - 10 1122
          AK298496.1 - 10 1001
          AK299362.1 - 10 864
          BC003576.1 - 10 1158 3547017
          BC015766.1 - 10 1033 4856734
          BT007207.1 - 10 916
          BX248020.1 - 10 889
          DQ496098.1 - 10 1099
          EU716325.1 - 10 936
          FJ410030.1 - 10 936
          M95178.1 - 10 1027
          X15804.1 - 10 1114
          X55187.1 - 1 28

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          actin filament organization
          platelet activation
          platelet formation
          platelet morphogenesis
          regulation of apoptotic process
          focal adhesion assembly
          actin filament bundle assembly
          negative regulation of cellular component movement
          actin filament network formation
          actin crosslink formation
          platelet aggregation
          regulation of nucleic acid-templated transcription
          double-stranded RNA binding
          integrin binding
          calcium ion binding
          protein binding
          vinculin binding
          ligand-dependent nuclear receptor transcription coactivator activity
          protein homodimerization activity
          ion channel binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0