Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006437.3 | NP_006428.2 | 19, 20 | 2448 | ||
XM_011534931.1 | XP_011533233.1 | 18, 19 | 2498 | |||
XM_011534932.2 | XP_011533234.1 | 19, 20 | 2665 | |||
GenBank mRNA | AF057160.1 | - | 19, 20 | 2394 | ||
AF158255.1 | - | 19, 20 | 2448 | |||
AK304399.1 | - | 19, 20 | 2416 | |||
D79999.2 | - | 18, 19 | 2347 |