Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127.3 | NP_001118.3 | 7 | 1008 | ||
NM_001166019.1 | NP_001159491.1 | 7 | 1008 | |||
NM_145730.2 | NP_663782.2 | 7 | 1008 | |||
GenBank mRNA | BC046242.1 | - | 7 | 1008 | 5929327 | |
CT841508.1 | - | 6 | 851 | |||
L13939.1 | - | 6 | 867 |