Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AKR1B1 SIRNA Assays ›
          Gene Symbol:
          AKR1B1
          Gene Name:
          aldo-keto reductase family 1 member B
          Gene Aliases:
          ADR, ALDR1, ALR2, AR
          Chromosome Location:
          Chr.7: 134442352 - 134459136 on Build GRCh38
          UniGene ID:
          Hs.521212
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS100407
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AKR1B1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001628.2 NP_001619.1 5 579
          XM_005250234.3 XP_005250291.1 5 588
          GenBank mRNA AB209796.1 - 5 498
          AF328729.1 - 5 520
          AK313439.1 - 5 570
          BC000260.1 - 5 579 3357652
          BC005387.1 - 5 543 3960811
          BC010391.1 - 5 545 4049736
          BT019859.1 - 5 505
          CR542203.1 - 5 505
          J04795.1 - 5 514
          J05017.1 - 5 541
          J05474.1 - 5 537
          M34720.1 - 5 518
          X15414.1 - 5 550

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          carbohydrate metabolic process
          sorbitol biosynthetic process
          C21-steroid hormone biosynthetic process
          response to stress
          response to water deprivation
          naphthalene metabolic process
          stress-activated protein kinase signaling cascade
          norepinephrine metabolic process
          daunorubicin metabolic process
          doxorubicin metabolic process
          fructose biosynthetic process
          positive regulation of JAK-STAT cascade
          positive regulation of smooth muscle cell proliferation
          oxidation-reduction process
          maternal process involved in female pregnancy
          cellular response to hydrogen peroxide
          inner medullary collecting duct development
          response to thyroid hormone
          cellular response to methylglyoxal
          cellular response to peptide
          alditol:NADP+ 1-oxidoreductase activity
          aldo-keto reductase (NADP) activity
          electron carrier activity
          glyceraldehyde oxidoreductase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jwk2j:80/100.66.76.207:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0